Anti-TMUB1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-7664R-CY5.5
10466-646EA 792.04 USD
10466-646
Anti-TMUB1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
May contribute to the regulation of translation during cell-cycle progression. May contribute to the regulation of cell proliferation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TMUB1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR