Anti-DNASE1L3 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-7653R-CY5.5
10466-662EA 792.04 USD
10466-662
Anti-DNASE1L3 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Has DNA hydrolytic activity. Does not bind to actin. Cleaves chromatin DNA to nucleosomal units.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DNASE1L3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR