Anti-DNASE2 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-7652R-CY3
10467-266EA 792.05 USD
10467-266
Anti-DNASE2 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Hydrolyzes DNA under acidic conditions with a preference for double-stranded DNA. Plays a major role in the degradation of nuclear DNA in cellular apoptosis during development. Necessary for proper fetal development and for definitive erythropoiesis in fetal liver, where it degrades nuclear DNA expelled from erythroid precursor cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DNASE2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR