Anti-ACSM3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-7641R-HRP
10466-354EA 792.04 USD
10466-354
Anti-ACSM3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
ACSM3

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: ACSM3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR