Anti-ACSM3 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-7641R-CY3
10466-344EA 792.04 USD
10466-344
Anti-ACSM3 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
ACSM3

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ACSM3
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR