Anti-SLC14A1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-7639R-CY3
10466-296EA 792.04 USD
10466-296
Anti-SLC14A1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
SLC14A1 is one of two major mammalian urea transporters, which play a critical role in the urine-concentrating mechanism. Their abundance is regulated by vasopressin, glucocorticoids, and mineralocorticoids. These regulatory mechanisms may be important in disease states such as diabetes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC14A1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR