Anti-SLC14A1 Rabbit Polyclonal Antibody (Biotin)

BS-7639R-BIO
10466-294EA 529.29 USD
10466-294
Anti-SLC14A1 Rabbit Polyclonal Antibody (Biotin)
SLC14A1 is one of two major mammalian urea transporters, which play a critical role in the urine-concentrating mechanism. Their abundance is regulated by vasopressin, glucocorticoids, and mineralocorticoids. These regulatory mechanisms may be important in disease states such as diabetes.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: SLC14A1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR