Anti-RHCG Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-7638R-CY3
10466-272EA 792.04 USD
10466-272
Anti-RHCG Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Functions as an electroneutral and bidirectional ammonium transporter. May regulate transepithelial ammonia secretion.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RHCG
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR