Anti-MTFR1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-7632R-CY5.5
10466-204EA 792.04 USD
10466-204
Anti-MTFR1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
This gene encodes a mitochondrial protein that is characterized by a poly-proline rich region. A chicken homolog of this protein promotes mitochondrial fission and the mouse homolog protects cells from oxidative stress. A related pseudogene of this gene is found on chromosome X.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MTFR1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR