Anti-MTCH1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-7630R-FITC
10466-160EA 776.97 USD
10466-160
Anti-MTCH1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Potential mitochondrial transporter. May play a role in apoptosis.Tissue specificity: Widely expressed with a predominant expression in brain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MTCH1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR