Anti-MTCH1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-7630R-CY3
10466-152EA 746.39 USD
10466-152
Anti-MTCH1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Potential mitochondrial transporter. May play a role in apoptosis.Tissue specificity: Widely expressed with a predominant expression in brain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MTCH1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR