Anti-MFF Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-7628R-A350
10466-094EA 792.04 USD
10466-094
Anti-MFF Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
MFF belongs to the tango11 family and plays a role in mitochondrial and peroxisomal fission. It is highly expressed in heart, kidney, liver, brain, muscle and stomach. There are five named isoforms.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MFF
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR