Anti-HAX1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-7626R-A350
10466-046EA 792.04 USD
10466-046
Anti-HAX1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
RelevanceHAX1 associates with HS1, binding to its N-terminal region. It is also known to associate with PKD2 (involved in polycystic kidney disease) and with cortactin/EMS1. HAX1 is also reported to bind to hairpin structures in vimentin and DNA polymerase beta mRNAs, so may play a role in mRNA stability and transport. It may also function in promoting cell survival. Defects in HAX1 are the cause of autosomal recessive severe congenital neutropenia 3 (SCN3) also called Kostmann disease.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HAX1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR