Anti-DUSP2 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-7609R-CY7
10465-732EA 792.04 USD
10465-732
Anti-DUSP2 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
DUSP2 is a dual-specific thr/tyr phosphatase that is a physiologically relevant MAP kinase phosphatase. The gene is located on 2q11.2-q11 and contains 4 exons that span approximately 2.3kb. Under specific stress conditions, p53 regulates transcription of PAC1 through a new p53-binding site, and that PAC1 is necessary and sufficient for p53-mediated apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DUSP2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR