Anti-IKBIP Rabbit Polyclonal Antibody (Biotin)

BS-7600R-BIO
10465-514EA 529.29 USD
10465-514
Anti-IKBIP Rabbit Polyclonal Antibody (Biotin)
Target of p53/TP53 with pro-apoptotic function.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: IKBIP
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR