Anti-IKBIP Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-7600R-A350
10465-506EA 792.04 USD
10465-506
Anti-IKBIP Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Target of p53/TP53 with pro-apoptotic function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IKBIP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR