Anti-DSTYK Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-7597R-CY5
10465-470EA 791.8 USD
10465-470
Anti-DSTYK Rabbit Polyclonal Antibody (Cy5®)
Antibodies
The RIPK5 gene encodes a dual serine/threonine and tyrosine protein kinase that is expressed in multiple tissues. Multiple alternatively spliced transcript variants have been found, but the biological validity of some variants has not been determined.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DSTYK
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR