Anti-GLA Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7593R
10465-360EA 621.96 USD
10465-360
Anti-GLA Rabbit Polyclonal Antibody
Antibodies
Galactosidase alpha is involved in the hydrolysis of terminal, non reducing alpha D galactose residues in alpha D galactosides, including galactose oligosaccharides, galactomannans and galactohydrolase. Defects in GLA are the cause of Fabry's disease (FD). FD is a rare X-linked sphingolipidosis disease where glycolipid accumulates in many tissues. Clinical recognition in males results from characteristic skin lesions (angiokeratomas) over the lower trunk. Patients may show ocular deposits, febrile episodes, and burning pain in the extremities. Death results from renal failure, cardiac or cerebral complications of hypertension or other vascular disease. Heterozygous females may exhibit the disorder in an attenuated form, they are more likely to show corneal opacities.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: Galactosidase alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR