Anti-A4GALT Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-7588R-A555
10465-276EA 792.04 USD
10465-276
Anti-A4GALT Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Necessary for the biosynthesis of the Pk antigen of blood histogroup P. Catalyzes the transfer of galactose to lactosylceramide and galactosylceramide. Necessary for the synthesis of the receptor for bacterial verotoxins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: A4GALT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR