Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BMF Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-7587R-A488
The BH3-only proteins, which include Blk, Bad, Bik, Hrk, BID, Bim, NOXA, PUMA and Bmf, are proapoptotic members of the Bcl-2 family. Bcl-2 modifying factor (Bmf) is a BH3-only protein that binds prosurvival Bcl-2 family members to initiate apoptosis. Bmf is sequestered to Myosin V motors on Actin in the cytoskeleton by associating with Dynein light chain 2 (DLC2) homodimers. If the cell undergoes loss of attachment (anoikis), the cytoskeleton is disrupted and Bmf is released from DLC2. Bmf then translocates to the mitochondria, where Bcl-2 (an anti-apoptotic family member) is sequestered. The BH3 domain of Bmf facilitates binding to a hydrophobic groove on the surface of Bcl-2. Binding results in a caspase cascade leading to apoptosis. Bmf is widely expressed in tissues such as pancreas, liver and kidney, and in hematopoietic tissues. The gene encoding Bmf maps to chromosome 15q14.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BMF
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-7587R-A488 10465-250 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol BMF
Antigen Name Bcl2 modifying Factor
Reactivity Rat, Human, Mouse
Conjugation Alexa Fluor® 488
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Gene ID 90427
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at -20°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty