Anti-BCL2L14 Rabbit Polyclonal Antibody (Biotin)

BS-7585R-BIO
10465-208EA 529.29 USD
10465-208
Anti-BCL2L14 Rabbit Polyclonal Antibody (Biotin)
Plays a role in apoptosis.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: BCL2L14
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR