Anti-BCL2L14 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-7585R-A488
10465-202EA 792.04 USD
10465-202
Anti-BCL2L14 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Plays a role in apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BCL2L14
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR