Anti-BCL2L15 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7582R
10464-894EA 621.96 USD
10464-894
Anti-BCL2L15 Rabbit Polyclonal Antibody
Antibodies
BCL2L15

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: BCL2L15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR