Anti-ZHX2 Rabbit Polyclonal Antibody (Biotin)

BS-7581R-BIO
10464-880EA 529.29 USD
10464-880
Anti-ZHX2 Rabbit Polyclonal Antibody (Biotin)
Acts as a transcriptional repressor. Represses the promoter activity of the CDC25C gene stimulated by NFYA.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: ZHX2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR