Anti-ZHX2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-7581R-A350
10464-872EA 792.04 USD
10464-872
Anti-ZHX2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Acts as a transcriptional repressor. Represses the promoter activity of the CDC25C gene stimulated by NFYA.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZHX2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR