Anti-FEM1B Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-7113R-CY3
10461-992EA 792.04 USD
10461-992
Anti-FEM1B Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Component of an E3 ubiquitin-protein ligase complex, in which it may act as a substrate recognition subunit. Involved in apoptosis by acting as a death receptor-associated protein that mediates apoptosis. Also involved in glucose homeostasis in pancreatic islet. Functions as an adapter/mediator in replication stress-induced signaling that leads to the activation of CHEK1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FEM 1B
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR