Anti-EDA2R Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-7111R-CY7
10461-950EA 792.04 USD
10461-950
Anti-EDA2R Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Receptor for EDA isoform A2, but not for EDA isoform A1. Mediates the activation of the NF-kappa-B and JNK pathways. Activation seems to be mediated by binding to TRAF3 and TRAF6.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EDA2R
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR