Anti-SCGB1D2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-6546R-A555
10449-474EA 791.8 USD
10449-474
Anti-SCGB1D2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
The protein encoded by this gene is a member of the lipophilin subfamily, part of the uteroglobin superfamily, and is an ortholog of prostatein, the major secretory glycoprotein of the rat ventral prostate gland. Lipophilin gene products are widely expressed in normal tissues, especially in endocrine-responsive organs. Assuming that human lipophilins are the functional counterparts of prostatein, they may be transcriptionally regulated by steroid hormones, with the ability to bind androgens, other steroids and possibly bind and concentrate estramustine, a chemotherapeutic agent widely used for prostate cancer. Although the gene has been reported to be on chromosome 10, this sequence appears to be from a cluster of genes on chromosome 11 that includes mammaglobin 2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCGB1D2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR