Anti-LDOC1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-6543R-CY5
10449-434EA 792.04 USD
10449-434
Anti-LDOC1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
May have an important role in the development and/or progression of some cancers.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LDOC1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR