Anti-HSPA6 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-6540R-A647
10449-356EA 792.04 USD
10449-356
Anti-HSPA6 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
In cooperation with other chaperones, Hsp70s stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage. Sequence similarities Belongs to the heat shock protein 70 family.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HSPA6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR