Anti-HOXB2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-6538R-A555
10448-938EA 791.8 USD
10448-938
Anti-HOXB2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HOXB2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR