Anti-GCSAM Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-6536R-CY7
10449-254EA 792.07 USD
10449-254
Anti-GCSAM Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Involved in the negative regulation of lymphocyte motility. It mediates the migration-inhibitory effects of IL6. Serves as a positive regulator of the RhoA signaling pathway. Enhancement of RhoA activation results in inhibition of lymphocyte and lymphoma cell motility by activation of its downstream effector ROCK. Is a regulator of B-cell receptor signaling, that acts through SYK kinase activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GCET2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR