Anti-KIAA1324 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-6533R-CY5.5
10449-178EA 792.04 USD
10449-178
Anti-KIAA1324 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Estrogen-induced gene 121 protein may play a role as a marker of hyperestrogenic state and estrogen-related type I endometrial carcinoma. It belongs to the UPF0577 family.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KIAA1324
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR