Anti-KIAA1324 Rabbit Polyclonal Antibody (Biotin)

BS-6533R-BIO
10449-172EA 529.29 USD
10449-172
Anti-KIAA1324 Rabbit Polyclonal Antibody (Biotin)
Estrogen-induced gene 121 protein may play a role as a marker of hyperestrogenic state and estrogen-related type I endometrial carcinoma. It belongs to the UPF0577 family.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: KIAA1324
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR