Anti-CKMT1B Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-6522R-A488
10448-982EA 791.8 USD
10448-982
Anti-CKMT1B Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Reversibly catalyzes the transfer of phosphate between ATP and various phosphogens (e.g. creatine phosphate). Creatine kinase isoenzymes play a central role in energy transduction in tissues with large, fluctuating energy demands, such as skeletal muscle, heart, brain and spermatozoa.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CKMT1B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR