Anti-CDK5RAP3 Rabbit Polyclonal Antibody (Biotin)

BS-6518R-BIO
10448-834EA 529.29 USD
10448-834
Anti-CDK5RAP3 Rabbit Polyclonal Antibody (Biotin)
Neuronal CDC2-like kinase, which is involved in the regulation of neuronal differentiation, is composed of a catalytic subunit, CDK5, and an activating subunit, p25NCK5A. The protein encoded by this gene binds to p25NCK5A and therefore may be involved in neuronal differentiation. The encoded protein, which may be a substrate of neuronal CDC2-like kinase, has also been found in vascular endothelial cells, where it mediates cell proliferation.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CDK5RAP3
Clonality: Polyclonal
Clone: 0
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR