Anti-CA8 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-6516R-CY7
10448-794EA 792.04 USD
10448-794
Anti-CA8 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
The protein encoded by this gene was initially named CA-related protein because of sequence similarity to other known carbonic anhydrase genes. However, the gene product lacks carbonic anhydrase activity (i.e., the reversible hydration of carbon dioxide). The gene product continues to carry a carbonic anhydrase designation based on clear sequence identity to other members of the carbonic anhydrase gene family. The absence of CA8 gene transcription in the cerebellum of the lurcher mutant in mice with a neurologic defect suggests an important role for this acatalytic form.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CA8
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR