Anti-ABCA2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-6515R-A488
10448-756EA 792.04 USD
10448-756
Anti-ABCA2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
C17orf71 belongs to the UPF0487 family. There are 3 isoforms produced by alternative splicing.Positive control;Human Anaplastic Thyroid Carcinoma, Human Breast Carcinoma.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ABCA2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR