Anti-A2BP1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-6513R-A488
10448-556EA 792.04 USD
10448-556
Anti-A2BP1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
RNA-binding protein that regulates alternative splicing events by binding to 5'-UGCAUGU-3' elements. Regulates alternative splicing of tissue-specific exons and of differentially spliced exons during erythropoiesis.Predominantly expressed in muscle and brain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: A2BP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR