Anti-CPE Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-6478R-CY3
10447-822EA 791.8 USD
10447-822
Anti-CPE Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Removes residual C-terminal Arg or Lys remaining after initial endoprotease cleavage during prohormone processing. Processes proinsulin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CPE
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR