Anti-XAB2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-6080R-A350
10430-078EA 792.04 USD
10430-078
Anti-XAB2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
XAB2 protein (XPA binding protein 2) was first identified through its interaction with XPA, a key factor in nucleotide excision repair pathways. XAB2 therefore appears to be involved in transcription coupled DNA repair and transcription. Immunoprecipitation experiments have also demonstrated that a fraction of XAB2 interacts with the transcription coupled repair specific proteins CSA and CSB as well as with RNA polymerase II. Microinjection experiments with XAB2 resulted in the inhibition of both transcription coupled repair and transcription.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: XAB2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR