Anti-Gene 21 protein/NPRL2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-6079R-A555
10430-058EA 792.04 USD
10430-058
Anti-Gene 21 protein/NPRL2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
NPR2L is homologous to yeast nitrogen permease and is a candidate tumor suppressor, being a negative regulator of cell cycle. Most abundant in skeletal muscle, followed by brain, liver, and pancreas, with lower amounts in lung, kidney, placenta, and heart. Expressed in most lung cancer cell lines tested. There are two isoforms, produced by alternative splicing.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Gene 21 protein/NPRL2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR