Anti-NIT1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-6077R-A647
10429-596EA 791.8 USD
10429-596
Anti-NIT1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Play a role in cell growth and apoptosis: loss of expression promotes cell growth and resistance to DNA damage stress. Has tumor suppressor properties that enhances the apoptotic responsiveness in cancer cells; this effect is additive to the tumor suppressor activity of FHIT. it is also a negative regulator of primary T-cells. Has apparently no omega-amidase activity such as NIT2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NIT1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR