Anti-MTUS1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-6076R-CY7
10429-982EA 791.8 USD
10429-982
Anti-MTUS1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Cooperates with AGTR2 to inhibit ERK2 activation and cell proliferation. May be required for AGTR2 cell surface expression. Together with PTPN6, induces UBE2V2 expression upon angiotensin-II stimulation. Isoform 1 inhibits breast cancer cell proliferation, delays the progression of mitosis by prolonging metaphase and reduces tumor growth.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MTUS1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR