Anti-MTSS1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-6075R-CY5.5
10429-796EA 792.05 USD
10429-796
Anti-MTSS1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
May be related to cancer progression or tumor metastasis in a variety of organ sites, most likely through an interaction with the actin cytoskeleton.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MTSS1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR