Anti-LRP12 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-6071R-A488
10429-894EA 792.04 USD
10429-894
Anti-LRP12 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Probable receptor, which may be involved in the internalization of lipophilic molecules and/or signal transduction. May act as a tumor suppressor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LRP12
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR