Anti-P3H3 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-6068R-CY7
10429-836EA 792.04 USD
10429-836
Anti-P3H3 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
LEPREL2 belongs to a family of collagen prolyl hydroxylases required for proper collagen biosynthesis, folding, and assembly.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: P3H3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR