Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-P3H3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-6068R-A555
10429-584
Anti-P3H3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
LEPREL2 belongs to a family of collagen prolyl hydroxylases required for proper collagen biosynthesis, folding, and assembly.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: P3H3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty