Anti-PPIA Rabbit Polyclonal Antibody (Biotin)

BS-5912R-BIO
10426-386EA 545.21 USD
10426-386
Anti-PPIA Rabbit Polyclonal Antibody (Biotin)
PPIases accelerate the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: PPIA
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR