Anti-DLD Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-5909R-A350
10426-304EA 746.42 USD
10426-304
Anti-DLD Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
DeltaD is one of the 4 members of the zebrafish Delta family of Notch ligands, important in the control of cell differentiation in many tissues.DeltaD acts as a ligand for Notch receptors and is involved in primary neurogenesis and somitogenesis. It can activate Notch receptors, thereby playing a key role in lateral inhibition, a process that prevents the immediate neighbors of each nascent neural cell from simultaneously embarking on neural differentiation. It is required in somite segmentation to keep the oscillations of neighboring presomitic mesoderm cells synchronized.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DLD
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR